Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134188.3 | NP_598949.3 | 3 | 1728 | ||
GenBank mRNA | AK141651.1 | - | 3 | 1715 | ||
AK172617.1 | - | 3 | 1724 | |||
BC064469.1 | - | 3 | 1700 | 6508650 |