Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134189.2 | NP_598950.2 | 3 | 467 | ||
GenBank mRNA | AK033515.1 | - | 2 | 125 | ||
AK131155.1 | - | 3 | 445 | |||
AK143714.1 | - | 3 | 465 | |||
AK159362.1 | - | 3 | 465 | |||
AK162424.1 | - | 3 | 467 | |||
AK172609.1 | - | 3 | 447 | |||
BB626609.1 | - | 3 | 467 | |||
BC060617.1 | - | 3 | 450 | 6836791 |