Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134192.3 | NP_598953.1 | 1 | 831 | ||
GenBank mRNA | AF394950.1 | - | 1 | 830 | ||
BC134400.1 | - | 1 | 737 |