Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PCDH7 SIRNA Assays ›
Gene Symbol:
PCDH7
Gene Name:
protocadherin 7
Gene Aliases:
BH-Pcdh, BHPCDH, PPP1R120
Chromosome Location:
Chr.4: 30720378 - 31146801 on Build GRCh38
UniGene ID:
Hs.479439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s10110
Validated
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
1 umol
10 umol
Availability
Inventoried
Catalog #
4390824
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PCDH7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173523.1
NP_001166994.1
1
2216
NM_002589.2
NP_002580.2
1
2216
NM_032456.2
NP_115832.1
1
2223
NM_032457.3
NP_115833.2
1
2216
XM_005248163.3
XP_005248220.1
1
2251
XM_005248164.3
XP_005248221.1
1
2251
XM_005248166.3
XP_005248223.1
1
2250
XM_011513842.2
XP_011512144.1
1
2252
XM_011513843.2
XP_011512145.1
1
2251
XM_011513844.2
XP_011512146.1
1
2252
XM_017008259.1
XP_016863748.1
1
2252
XM_017008260.1
XP_016863749.1
1
2252
XM_017008261.1
XP_016863750.1
1
2252
XM_017008262.1
XP_016863751.1
1
2252
XM_017008263.1
XP_016863752.1
1
2253
XM_017008264.1
XP_016863753.1
1
2253
XM_017008265.1
XP_016863754.1
1
2253
XM_017008266.1
XP_016863755.1
1
2253
XM_017008267.1
XP_016863756.1
1
2253
XM_017008268.1
XP_016863757.1
1
2253
XM_017008269.1
XP_016863758.1
1
2253
XM_017008270.1
XP_016863759.1
1
2252
XM_017008271.1
XP_016863760.1
1
2252
XM_017008272.1
XP_016863761.1
1
2251
GenBank mRNA
AB006755.1
-
1
2218
AB006756.1
-
1
1775
AB006757.1
-
2
1922
Back To Top
More Information
Matching Taqman Gene Expression Assay:
Hs00159908_m1
Panther Classification:
Molecular Function -
cadherin
Gene Ontology Categories:
Function(s)
Process(es)
platelet degranulation
homophilic cell adhesion via plasma membrane adhesion molecules
calcium ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-qvdz4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0