Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Havcr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134250.2 | NP_599011.2 | 7 | 848 | ||
GenBank mRNA | AF399831.1 | - | 7 | 794 | ||
AF450241.1 | - | 7 | 844 | |||
AK148474.1 | - | 7 | 849 | |||
AK170775.1 | - | 8 | 669 | |||
AY553334.1 | - | 7 | 841 | |||
BC106851.2 | - | 7 | 806 | 40046221 | ||
BC106852.2 | - | 7 | 806 | 40046222 |