Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc12a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083902.1 | NP_001077371.1 | 8 | 1076 | ||
NM_134251.2 | NP_599012.2 | 9 | 1192 | |||
GenBank mRNA | AF319951.1 | - | 9 | 1186 | ||
AK087781.1 | - | 5 | 782 | |||
AK165080.1 | - | 4 | 699 | |||
BC030926.1 | - | 8 | 1076 | 4910321 |