Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172776.2 | NP_766364.1 | 6 | 1180 | ||
GenBank mRNA | AF536195.1 | - | 6 | 1252 | ||
AF536196.1 | - | 7 | 988 | |||
AK052597.1 | - | 6 | 1181 | |||
BC126952.1 | - | 7 | 993 | 40090863 |
Gene Symbol: Slc22a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134256.1 | NP_599017.1 | 7 | 1121 | ||
GenBank mRNA | AB056442.1 | - | 7 | 1121 | ||
AB056443.1 | - | 5 | 770 |