Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134256.1 | NP_599017.1 | 8 | 1299 | ||
GenBank mRNA | AB056442.1 | - | 8 | 1299 | ||
AB056443.1 | - | 6 | 948 |
Gene Symbol: Slc22a29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172776.2 | NP_766364.1 | 7 | 1358 | ||
GenBank mRNA | AF536195.1 | - | 7 | 1430 | ||
AF536196.1 | - | 8 | 1166 | |||
AK052597.1 | - | 7 | 1359 | |||
BC126952.1 | - | 8 | 1171 | 40090863 |