Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd99l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199349.1 | NP_001186278.1 | 6 | 670 | ||
NM_138309.3 | NP_612182.1 | 5 | 601 | |||
GenBank mRNA | AK031264.1 | - | 6 | 427 | ||
AK080225.1 | - | 6 | 426 | |||
AK166275.1 | - | 6 | 421 | |||
AY078163.1 | - | 5 | 397 | |||
BC031736.1 | - | 5 | 376 | 4013657 | ||
EF516990.1 | - | 6 | 428 |