Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1foo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138311.2 | NP_612184.1 | 2, 3 | 405 | ||
GenBank mRNA | AK135944.1 | - | 2, 3 | 409 | ||
AK162137.1 | - | 2, 3 | 384 | |||
AK162154.1 | - | 2, 3 | 402 | |||
AK165127.1 | - | 3, 4 | 487 | |||
AY007195.1 | - | 2, 3 | 405 | |||
BC137916.1 | - | 2, 3 | 401 | 8860935 | ||
BC145046.1 | - | 2, 3 | 401 |