Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138313.3 | NP_612186.2 | 3, 4 | 560 | ||
GenBank mRNA | AK040622.1 | - | 3, 4 | 517 | ||
AK084658.1 | - | 3, 4 | 570 | |||
AK169903.1 | - | 3, 4 | 517 | |||
AY029253.1 | - | 1, 2 | 286 | |||
BC079650.1 | - | 3, 4 | 574 | 6847314 |