Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Socs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138657.3 | NP_619598.1 | 6 | 1352 | ||
GenBank mRNA | AF424814.1 | - | 6 | 1353 | ||
AK147580.1 | - | 6 | 1450 | |||
AK147650.1 | - | 6 | 1450 |