Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdha12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138663.2 | NP_619604.1 | 1 | 627 | ||
GenBank mRNA | AB008180.1 | - | 1 | 520 | ||
AY013759.1 | - | 1 | 627 |