Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ufsp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138668.2 | NP_619609.1 | 4 | 347 | ||
GenBank mRNA | AK075795.1 | - | 4 | 342 | ||
AK078309.1 | - | 4 | 348 | |||
AK160164.1 | - | 3 | 422 | |||
BC005503.1 | - | 4 | 348 | 3490744 |