Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam195b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033231.2 | NP_001028403.1 | 3 | 227 | ||
GenBank mRNA | AK147780.1 | - | 3 | 205 | ||
BC002190.1 | - | 2 | 159 | |||
BC003940.1 | - | 2 | 159 | |||
BC025536.1 | - | 3 | 200 | |||
BC052229.1 | - | 2 | 329 | |||
BI987638.1 | - | 3 | 227 |