Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018073.2 | NP_001018083.2 | 2 | 416 | ||
NM_001308054.1 | NP_001294983.1 | 2 | 677 | |||
NM_004563.3 | NP_004554.3 | 2 | 416 | |||
XM_006720158.2 | XP_006720221.1 | 2 | 409 | |||
GenBank mRNA | AK129934.1 | - | 2 | 234 | ||
AK301400.1 | - | 2 | 260 | |||
AK307913.1 | - | 2 | 645 | |||
AK311710.1 | - | 2 | 647 | |||
BC001454.2 | - | 2 | 206 | 3138368 | ||
BQ957219.1 | - | 2 | 634 | |||
BX248272.1 | - | 2 | 190 | |||
CR456913.1 | - | 2 | 141 | |||
DB028389.1 | - | 2 | 409 | |||
HY022221.1 | - | 3 | 316 | |||
KU178243.1 | - | 2 | 141 | |||
KU178244.1 | - | 2 | 141 | |||
X92720.1 | - | 2 | 207 |