Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138751.2 | NP_620090.1 | 1 | 438 | ||
GenBank mRNA | AB041540.1 | - | 1 | 371 | ||
AK030761.1 | - | 1 | 367 | |||
AK031346.1 | - | 1 | 388 | |||
AK076147.1 | - | 1 | 377 | |||
AK133790.1 | - | 1 | 375 | |||
AK162304.1 | - | 1 | 376 | |||
BC019751.1 | - | 1 | 349 | 4020610 |