Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019424.2 | NP_062297.1 | 14 | 1564 | ||
GenBank mRNA | AK031397.1 | - | 14 | 1578 | ||
AK053974.1 | - | 12 | 1397 | |||
AK154994.1 | - | 14 | 1753 | |||
AK171225.1 | - | 14 | 1540 | |||
AK211068.1 | - | 2 | 107 | |||
BC046329.1 | - | 12 | 1385 | |||
BC058332.1 | - | 14 | 1547 | |||
BC089331.1 | - | 14 | 1547 | 6848298 | ||
U78315.1 | - | 13 | 1499 | |||
U97149.1 | - | 13 | 1513 |