Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133792.2 | NP_598553.1 | 2 | 339 | ||
GenBank mRNA | AF468958.1 | - | 2 | 251 | ||
AK028614.1 | - | 2 | 339 | |||
AK085194.1 | - | 2 | 328 | |||
AK155004.1 | - | 2 | 335 | |||
AK163111.1 | - | 2 | 328 | |||
AK170814.1 | - | 2 | 327 | |||
AK180645.1 | - | 1 | 146 | |||
AK186155.1 | - | 2 | 148 | |||
AK188333.1 | - | 2 | 146 | |||
AK189283.1 | - | 1 | 146 | |||
AK203519.1 | - | 1 | 132 | |||
AK205252.1 | - | 2 | 146 | |||
AK205480.1 | - | 2 | 146 | |||
AK207442.1 | - | 2 | 146 | |||
AK211077.1 | - | 2 | 146 | |||
AK212406.1 | - | 2 | 146 | |||
AK212507.1 | - | 2 | 146 | |||
AK212587.1 | - | 2 | 146 | |||
AK212623.1 | - | 2 | 146 | |||
AK214459.1 | - | 1 | 94 | |||
AK215050.1 | - | 2 | 146 | |||
AK215274.1 | - | 2 | 146 | |||
AK215692.1 | - | 2 | 146 | |||
AK215942.1 | - | 2 | 148 | |||
AK217196.1 | - | 2 | 146 | |||
AK219754.1 | - | 2 | 146 | |||
AK220071.1 | - | 2 | 146 | |||
BC019373.1 | - | 2 | 339 | 3964729 |