Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138952.3 | NP_620402.1 | 4 | 730 | ||
GenBank mRNA | AF461040.1 | - | 4 | 628 | ||
AF487539.1 | - | 4 | 628 | |||
AK159593.1 | - | 4 | 754 | |||
AK172319.1 | - | 4 | 752 | |||
AK172417.1 | - | 4 | 752 | |||
BC025611.1 | - | 2 | 248 | |||
BC050845.1 | - | 4 | 743 | |||
BC058088.1 | - | 4 | 875 | |||
BC069878.1 | - | 4 | 755 | 5316066 |