Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138955.3 | NP_620405.3 | 3 | 571 | ||
GenBank mRNA | AF378330.1 | - | 2 | 402 | ||
AF411084.1 | - | 3 | 449 | |||
AF425077.1 | - | 3 | 451 | |||
AJ426047.2 | - | 3 | 450 | |||
AK043147.1 | - | 3 | 572 | |||
AK138415.1 | - | 2 | 412 | |||
AK157317.1 | - | 3 | 896 | |||
AY040865.1 | - | 3 | 415 | |||
BC016200.1 | - | 3 | 451 | 4511456 |