Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138955.3 | NP_620405.3 | 2 | 371 | ||
GenBank mRNA | AF378330.1 | - | 1 | 202 | ||
AF411084.1 | - | 2 | 249 | |||
AF425077.1 | - | 2 | 251 | |||
AJ426047.2 | - | 2 | 250 | |||
AK043147.1 | - | 2 | 372 | |||
AK138415.1 | - | 1 | 212 | |||
AK157317.1 | - | 2 | 696 | |||
AY040865.1 | - | 2 | 215 | |||
BC016200.1 | - | 2 | 251 | 4511456 |