Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001445.2 | NP_001001445.1 | 11 | 1909 | ||
GenBank mRNA | AB180097.1 | - | 10 | 1657 | ||
AJ620495.1 | - | 10 | 1697 | |||
AK051315.1 | - | 11 | 1767 | |||
AY445519.1 | - | 10 | 1657 | |||
AY452083.1 | - | 10 | 1690 | |||
AY452084.1 | - | 10 | 1660 |