Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177565.3 | NP_808233.1 | 2 | 331 | ||
GenBank mRNA | AK045753.1 | - | 2 | 332 | ||
BC096463.1 | - | 2 | 313 | 6466366 |