Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam65b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080381.1 | NP_001073850.1 | 14 | 1933 | ||
NM_001286100.1 | NP_001273029.1 | 14 | 1801 | |||
NM_001286101.1 | NP_001273030.1 | 14 | 1900 | |||
NM_029679.2 | NP_083955.1 | 14 | 1933 | |||
NM_178658.5 | NP_848773.3 | 13 | 1660 | |||
GenBank mRNA | AK031928.1 | - | 14 | 1886 | ||
AK036452.1 | - | 14 | 1880 | |||
AK036695.1 | - | 2 | 550 | |||
AK040887.1 | - | 13 | 1640 | |||
AK041665.1 | - | 13 | 1661 | |||
AK122269.1 | - | 14 | 1865 | |||
AK134538.1 | - | 14 | 1863 | |||
BC059179.1 | - | 14 | 1802 |
Gene Symbol: Ntrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | X17647.1 | - | 3511 |