Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp184 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183014.1 | NP_898835.1 | 6 | 1815 | ||
GenBank mRNA | AK036164.1 | - | 6 | 1831 | ||
BC053084.1 | - | 6 | 1815 | 5708591 |