Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198886.3 | NP_942589.3 | 2 | 532 | ||
GenBank mRNA | AK135305.1 | - | 1 | 431 | ||
BC020447.1 | - | 2 | 471 | |||
BC062159.1 | - | 2 | 532 | 30135322 |