Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290283.1 | NP_001277212.1 | 1 | 176 | ||
NM_183139.2 | NP_898962.2 | 1 | 176 | |||
GenBank mRNA | AK077214.1 | - | 1 | 138 | ||
AK139586.1 | - | 1 | 164 | |||
BC119245.1 | - | 1 | 176 | 8733994 | ||
BC145052.1 | - | 1 | 176 |