Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205068.1 | NP_001191997.1 | 6 | 1380 | ||
NM_178659.6 | NP_848774.1 | 6 | 1416 | |||
GenBank mRNA | AK032477.1 | - | 6 | 1304 | ||
AK037954.1 | - | 6 | 1415 | |||
AK038650.1 | - | 5 | 873 | |||
AK039116.1 | - | 6 | 1008 | |||
AK047586.1 | - | 6 | 1331 | |||
AK052144.1 | - | 6 | 1366 | |||
AK079104.1 | - | 6 | 1374 | |||
AK084669.1 | - | 6 | 1375 | |||
AK145394.1 | - | 6 | 1394 | |||
AK171919.1 | - | 6 | 1285 | |||
BC049148.1 | - | 6 | 1315 | |||
BC098198.1 | - | 5 | 3973 | |||
BC116707.1 | - | 6 | 1347 | 40096667 | ||
BC118020.1 | - | 6 | 1260 |