Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zzef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045536.2 | NP_001039001.1 | 50, 51 | 8186 | ||
GenBank mRNA | AK052734.1 | - | 2, 3 | 292 | ||
AK082066.1 | - | 4, 5 | 1347 | |||
BC026619.1 | - | 1, 2 | 1656 | |||
BC037463.1 | - | 1, 2 | 73 | |||
BC099976.1 | - | 10, 11 | 1641 |