Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170464.2 | NP_001163935.1 | 6, 7 | 1240 | ||
NM_002595.4 | NP_002586.2 | 6, 7 | 1240 | |||
XM_017019405.1 | XP_016874894.1 | 6, 7 | 682 | |||
XM_017019406.1 | XP_016874895.1 | 6, 7 | 697 | |||
XM_017019407.1 | XP_016874896.1 | 6, 7 | 1127 | |||
GenBank mRNA | AK290011.1 | - | 6, 7 | 1132 | ||
AK294698.1 | - | 5, 6 | 579 | |||
AK315214.1 | - | 6, 7 | 1150 | |||
BC033005.1 | - | 6, 7 | 729 | 4824359 | ||
X66360.1 | - | 6, 7 | 677 |