Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144785.2 | NP_659034.1 | 4 | 796 | ||
GenBank mRNA | AF529221.1 | - | 4 | 725 | ||
AK052752.1 | - | 4 | 766 | |||
BC021384.1 | - | 4 | 762 | 5064542 |