Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc22a19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144785.2 | NP_659034.1 | 2 | 586 | ||
GenBank mRNA | AF529221.1 | - | 2 | 515 | ||
AK052603.1 | - | 2 | 586 | |||
AK052752.1 | - | 2 | 556 | |||
BC021384.1 | - | 2 | 552 | 5064542 |