Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ggt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144786.2 | NP_659035.2 | 10 | 1346 | ||
GenBank mRNA | AK032051.1 | - | 10 | 1344 | ||
AK131175.1 | - | 7 | 3369 | |||
BC005772.1 | - | 10 | 1327 | 3602314 |