Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002596.3 | NP_002587.2 | 7 | 993 | ||
NM_212502.2 | NP_997667.1 | 7 | 911 | |||
NM_212503.2 | NP_997668.1 | 7 | 1001 | |||
XM_011509602.2 | XP_011507904.1 | 7 | 1051 | |||
XM_017001423.1 | XP_016856912.1 | 7 | 1146 | |||
GenBank mRNA | AB209307.1 | - | 5 | 1573 | ||
AK126879.1 | - | 4 | 997 | |||
AK293756.1 | - | 6 | 1275 | |||
AK295272.1 | - | 5 | 441 | |||
AK295654.1 | - | 6 | 1056 | |||
AK296320.1 | - | 7 | 959 | |||
AK308913.1 | - | 5 | 442 | |||
AY353237.1 | - | 7 | 826 | |||
AY353238.1 | - | 7 | 890 | |||
BC000281.2 | - | 1 | 78 | |||
BC011526.1 | - | 7 | 846 | 3855304 | ||
BT007299.1 | - | 6 | 643 | |||
X66362.1 | - | 4 | 367 |