Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb7c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145356.3 | NP_663331.1 | 4 | 1157 | ||
GenBank mRNA | AK045522.1 | - | 4 | 1121 | ||
AK156372.1 | - | 2 | 825 | |||
BC018187.1 | - | 2 | 854 | 4208891 | ||
BC070424.1 | - | 4 | 1157 | 6837447 |