Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172442.3 | NP_766030.3 | 9 | 2496 | ||
GenBank mRNA | AK086624.1 | - | 8 | 1065 | ||
BC044798.1 | - | 4 | 379 | |||
BC058647.1 | - | 9 | 1954 | 6405881 | ||
BC069975.1 | - | 8 | 1036 | |||
CN525673.1 | - | 2 | 184 |