Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172442.3 | NP_766030.3 | 4 | 1908 | ||
GenBank mRNA | AK086624.1 | - | 3 | 477 | ||
BC058647.1 | - | 4 | 1366 | 6405881 | ||
BC069975.1 | - | 3 | 448 | |||
CA328050.1 | - | 3 | 345 |