Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170599.2 | NP_733548.2 | 3 | 478 | ||
GenBank mRNA | AB079880.1 | - | 3 | 315 | ||
AK160146.1 | - | 3 | 441 | |||
BC057555.1 | - | 3 | 478 | 6416190 |