Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C2cd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174847.2 | NP_777272.2 | 6 | 1087 | ||
GenBank mRNA | AK017896.1 | - | 6 | 1072 | ||
AK077731.1 | - | 6 | 1048 | |||
AK080956.1 | - | 6 | 1031 | |||
AK086520.1 | - | 6 | 1089 | |||
AK143875.1 | - | 6 | 1061 | |||
AK169902.1 | - | 6 | 1023 | |||
AK220454.1 | - | 6 | 1072 | |||
BC043688.1 | - | 6 | 1048 | 5136661 |