Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smagp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033872.2 | NP_001029044.1 | 4 | 426 | ||
NM_001285413.1 | NP_001272342.1 | 3 | 409 | |||
NM_001285414.1 | NP_001272343.1 | 3 | 362 | |||
NM_174992.4 | NP_778157.2 | 4 | 616 | |||
GenBank mRNA | BC004728.1 | - | 4 | 417 | 3582955 | |
BC132460.1 | - | 3 | 223 | 40130737 | ||
BC132462.1 | - | 3 | 223 | 40130739 | ||
BE132795.1 | - | 4 | 282 | |||
BF018035.1 | - | 4 | 494 | |||
BY707988.1 | - | 4 | 351 | |||
CJ042753.1 | - | 4 | 379 | |||
CJ052870.1 | - | 3 | 304 | |||
CK129185.1 | - | 4 | 320 |