Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vcpkmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033236.2 | NP_001028408.2 | 2 | 389 | ||
GenBank mRNA | AK083148.1 | - | 1 | 468 | ||
AK144832.1 | - | 2 | 352 | |||
BC138942.1 | - | 2 | 471 | 8861964 | ||
BC138943.1 | - | 2 | 471 | 8861965 | ||
BC145471.1 | - | 2 | 471 |