Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pif1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172453.3 | NP_766041.1 | 1 | 781 | ||
GenBank mRNA | AK028460.1 | - | 1 | 782 | ||
AK049353.1 | - | 1 | 783 | |||
AK154072.1 | - | 1 | 794 | |||
AY100322.1 | - | 2 | 494 | |||
AY498715.1 | - | 1 | 781 | |||
BC046611.1 | - | 2 | 419 | 6396273 | ||
BC060108.1 | - | 1 | 772 |