Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Panx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172454.2 | NP_766042.2 | 4 | 1171 | ||
GenBank mRNA | AK028288.1 | - | 4 | 1138 | ||
AK036933.1 | - | 4 | 1127 | |||
BC052822.1 | - | 4 | 1139 | 30058169 |