Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003683.2 | NP_001003683.1 | 4 | 491 | ||
NM_001258312.1 | NP_001245241.1 | 4 | 518 | |||
NM_001258313.1 | NP_001245242.1 | 3 | 670 | |||
NM_001258314.1 | NP_001245243.1 | 2 | 347 | |||
NM_005019.4 | NP_005010.2 | 3 | 533 | |||
XM_011511323.2 | XP_011509625.1 | 3 | 310 | |||
XM_011511324.2 | XP_011509626.1 | 4 | 538 | |||
XM_011511325.2 | XP_011509627.1 | 3 | 665 | |||
XM_011511326.2 | XP_011509628.1 | 2 | 342 | |||
XM_017004294.1 | XP_016859783.1 | 3 | 533 | |||
XM_017004295.1 | XP_016859784.1 | 4 | 538 | |||
XM_017004296.1 | XP_016859785.1 | 4 | 460 | |||
XM_017004297.1 | XP_016859786.1 | 4 | 401 | |||
XM_017004298.1 | XP_016859787.1 | 4 | 583 | |||
XM_017004299.1 | XP_016859788.1 | 4 | 538 | |||
XM_017004300.1 | XP_016859789.1 | 4 | 583 | |||
XM_017004301.1 | XP_016859790.1 | 4 | 388 | |||
XM_017004302.1 | XP_016859791.1 | 3 | 665 | |||
GenBank mRNA | AB038228.1 | - | 2 | 224 | ||
AJ401610.1 | - | 3 | 252 | |||
AK294239.1 | - | 4 | 453 | |||
AK295657.1 | - | 4 | 323 | |||
AK301720.1 | - | 2 | 330 | |||
AL110263.1 | - | 3 | 644 | |||
BC022480.1 | - | 3 | 501 | 4814064 | ||
DA295167.1 | - | 4 | 541 | |||
DC396228.1 | - | 2 | 347 | |||
U40370.1 | - | 4 | 384 |