Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203319.1 | NP_976064.1 | 22 | 2961 | ||
GenBank mRNA | AK148257.1 | - | 22 | 3046 | ||
AK220399.1 | - | 19 | 2312 | |||
BC066077.1 | - | 22 | 2961 | 30354591 | ||
BC080298.1 | - | 22 | 2979 | 30362983 |