Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145361.2 | NP_663336.2 | 9 | 1979 | ||
GenBank mRNA | AK046604.1 | - | 7 | 827 | ||
AK194965.1 | - | 4 | 284 | |||
BC016566.1 | - | 5 | 525 | |||
BC039625.1 | - | 2 | 252 | |||
BC040692.1 | - | 2 | 252 | |||
BC055704.1 | - | 8 | 1046 | |||
BC069864.1 | - | 5 | 498 | |||
BC083320.1 | - | 8 | 952 | |||
BC115682.1 | - | 8 | 1039 | 40104217 | ||
BC115683.1 | - | 8 | 1039 | 40104219 |