Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145361.2 | NP_663336.2 | 9 | 2672 | ||
GenBank mRNA | AK046604.1 | - | 7 | 1520 | ||
BC016566.1 | - | 5 | 1218 | |||
BC039625.1 | - | 2 | 945 | |||
BC040692.1 | - | 2 | 945 | |||
BC055704.1 | - | 8 | 1739 | |||
BC069864.1 | - | 5 | 1191 | |||
BC083320.1 | - | 8 | 1645 |