Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mob3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172457.2 | NP_766045.2 | 2 | 210 | ||
GenBank mRNA | AK030481.1 | - | 2 | 246 | ||
AK170394.1 | - | 2 | 221 | |||
AK171197.1 | - | 234 | ||||
BC051170.1 | - | 1 | 199 | |||
BC058238.1 | - | 2 | 210 | 6307543 |