Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dot1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199322.1 | NP_955354.1 | 22 | 2829 | ||
GenBank mRNA | AK220271.1 | - | 22 | 2787 | ||
AY196089.1 | - | 22 | 2829 | |||
AY376663.1 | - | 19 | 2646 | |||
AY377920.1 | - | 15 | 1951 | |||
JF508521.1 | - | 22 | 2591 |